Whatman™ Grade 6 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
1006-110 1006-185 1006-090 1006-125 1006-240 1006-240 1006-070 1006-042 1006-185 1006-090 1006-125 1006-150 1006-042 1006-070 1006-150 1006-110
28462-170PK 62.34 CAD
28462-170 CA11008-516 CA11008-568 CA11008-570 CA11012-566 10035-236 10147-904 10147-902 10147-994 10147-988 10147-990 10147-992 CA10147-902 CA10147-904 CA10147-992 CA28462-170
Whatman™ Grade 6 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Papers Qualitative Filter Paper
A standard grade filter for boiler water analysis.
  • Twice as fast as Grade 5 with similar fine particle retention
  • Often specified for boiler water analysis applications
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.