8247 R95 Respirator, 3M™
Supplier: 3M
7100160544 7000002060
CA11006-300CS 907.14 CAD
CA11006-300 CA56621-932
8247 R95 Respirator, 3M™
Respirators Disposable Respirators Particulate Respirators
This respirator provides worker protection against certain oil and non-oil based aerosols including those with nuisance levels of organic vapors such as solvents, degreasers, and resins.
  • Protects against certain oil and non-oil based aerosols
  • If oil aerosols are present, respirator may be limited to 8 hours of continuous or intermittent use
  • Provides minimum filtration efficiency of 95%
(Nuisance level acid gas refers to concentrations not exceeding the OSHA PEL or applicable government occupational exposure limits, whichever is lower.) If oil aerosolsMore Product Information