KIMTECH PURE® M6 Face Mask, Kimberly-Clark Professional®
Supplier: Microflex
62477
CA10843-001CS 229.12 CAD
CA10843-001
KIMTECH PURE® M6 Face Mask, Kimberly-Clark Professional®
Masks Cleanroom Masks
Two-layer general-purpose mask with earloops provides comfort with BICOSOF® fabric inner facing.
  • Two layer, general purpose
  • Earloops
  • BICOSOF® fabric inner facing
Recommended for ISO Class 6 or higher cleanroom environments.
Packaging: Double-bagged.