HyClone™ HyCell TransFx-C Medium, HyClone products (Cytiva)
Supplier: Cytiva
SH30941.02 SH30942.01
CA10842-584EA 147.01 CAD
CA10842-584 CA76462-544
HyClone™ HyCell TransFx-C Medium, HyClone products (Cytiva)
Cell Culture Media
HyCell™ TransFx-C Media is a hydrolysate-free and regulatory-friendly media which supports transient transfection, growth, and expression of a variety of recombinant proteins in CHO cells.
  • Defined hydrolosate-free ADCF cell culture media formulation
  • Supports the transient expression of a variety of recombinant proteins
  • Same media for transient transfection and production
  • Developed through our Metabolic Pathway Design process
  • Component traceability
High-performing, animal-derived component-free (ADCF) cell culture media with sodium bicarbonate and without L-glutamine and poloxamer 188.
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO 9001 certified facilities.