Sentino® Microbiology Pump, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
13186 13186
10841-650EA 2581.72 CAD
10841-650 CA10841-650
Sentino® Microbiology Pump, Cytiva (Formerly Pall Lab)
Pumps Peristaltic Pumps
The Sentino® pump system is ideal for microbiological MF technique using Pall MicroFunnel™ filtration units. Designed to use peristaltic action to draw the sample through the filter and send filtrate directly to drain or waste collection. The housing is a PC ABS-blended polymer, the keypad and label PE, the pump head PEI with PTFE-filled PBT rollers. The fluid path adapter is PP.
  • Safety: The system eliminates the need for a fluid trap under vacuum, preventing implosion accidents
  • The disposable design also eliminates the need to clean and autoclave manifold
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable