Water, certified Molecular_Biology

Supplier: Cytiva

Synonyms: Hydrogen oxide

WFI-MGW-20L WFI-MGW-20L
10837-214EA 458.2 CAD
10837-214 CA10837-214
Water, certified Molecular_Biology
Water

Cytiva Molecular Grade water is quality tested and certified RNase, DNase, and protease free.


  • Purified utilizing a nine step process resulting in very high quality water


Every lot sent to our customer contains zero traces of toxic agents such as DEPC, and is well suited for use in diverse molecular biology applications.


Certifications: Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear Liquid (Visual)
TOC 500 ppb (USP 643)
Conductivity 5.0 µs/cm (USP 645)
DNAse None Detected
Protease None Detected
RNAse None Detected
GMP manufactured conform


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR