Uritainer™ 24 Hour Urine Collection Containers, Simport Scientific
Supplier: Simport Scientific
B360-25 B360-25 B360-35
CA10836-464CS 180.88 CAD
CA10836-464 76299-216 76299-218
Uritainer™ 24 Hour Urine Collection Containers, Simport Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
All models feature a unique snap-valve pour spout that is easily popped open to provide dripless pouring.
  • Unique snap-valve pour spout is easily popped open
  • Dripless pouring
  • Specially designed leakproof screw cap with liner for safer transport
  • Metal-free and resistant to hydrochloric acid
  • Large handle can be gripped comfortably
  • Volume level read in 50 ml increments
  • Can be gamma sterilized
This more conventionally styled urine bottle is available in 2.5 and 3.5 l sizes. It also incorporates some of the great features of the URISAFE® 24 Hour UrineMore Product Information