FiveGo™ F2 Portable pH Meters, METTLER TOLEDO®

Supplier: Mettler Toledo
30266889 30266882 30266881 30266946
CA10821-370EA 838.73 CAD
CA10821-370 CA10821-372 CA10821-374 CA10821-428
FiveGo™ F2 Portable pH Meters, METTLER TOLEDO®
Electrochemistry Meters pH and Multi-Parameter Meters
Ideally suited for mobile applications in the laboratory, at-line, or outdoors, the FiveGo™ portable instruments provide high quality pH and mV measurements with the simple click of a button. The robust and waterproof design makes the FiveGo™ instruments the perfect companion to get reliable measurement data even in harsh conditions.

  • Intuitive operation - Measurement made easy
  • Ergonomic design - Comfortable handling
  • Foldable foot stand provides optimal viewing while situated on a lab bench
  • Waterproof - Ideal for wet conditions
  • IP 67 waterproof protection
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range 0.00 … 14.00
pH resolution 0.01
pH accuracy ±0.01
pH calibration Max. 3 points, 4 predefined buffer groups
mV range −1,999 … 1,999 mV
mV resolution 1 mV
mV accuracy ±1 mV
Temperature range 0.0 … 100.0 °C (32.0 … 212.0 °F)
Temperature resolution 0.1 °C
Temperature accuracy ±0.5 °C
Temperature compensation Automatic or manual
Display 3.1" segmented LCD
Battery life >200 hrs
Power supply 4 × 1.5 V AAA LRO3 or 4 × 1.2 V AAA NiMH HR03 batteries
Data logging capacity 200 measurements, current calibration
IP rating IP67
Dimensions 188 × 77 × 33 mm
Connections BNC, Cinch (NTC 30 kΩ)
Alarm Acoustic/visual
Weight 0.26 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR