KIMBERLY-CLARK® Universal Precautions Lab Coats, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Microflex
10041 10043 10040 10044 10047 10042 10046
CA10815-732CS 614.76 CAD
CA10815-732 CA10815-736 CA10837-224 CA37001-670 CA67000-064 CA10815-734 10815-740
KIMBERLY-CLARK® Universal Precautions Lab Coats, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
Disposable, fluid-resistant lab coats are manufactured from a protective three-layer spunbond-meltblown-spunbond (SMS) fabric that offers exceptional resistance to blood strikethrough for moderate fluid contact
  • Disposable, fluid-resistant, three-layer SMS fabric
  • Six-snap front closure, three reinforced pockets, back vent, traditional collar, and knit cuffs
  • Tailored styling and fit enhances worker comfort
  • Non sterile, latex-free
Lab coats feature a six-snap front closure, three reinforced pockets, back vent, traditional collar, and knit cuffs. TailoredMore Product Information