Fi-Streem™ 2S Still 8 Liter Carboy, Labstrong Corporation
Supplier: LABSTRONG CORPORATION CA
A1052LS
CA10809-184EA 4073.75 CAD
CA10809-184
Fi-Streem™ 2S Still 8 Liter Carboy, Labstrong Corporation
Bottles Carboys
The LabStrong eight liters carboy allows for fully automatic operation when connected to Fi-Streem™ 2S stills.
  • Easy to grip handle on the top makes the carboy easy to lift
  • Rectangular base that occupies less space than a round carboy
  • Large opening at the top providing for easier filling and cleaning
  • Graduation marks
  • Dispense spigot
The product is a high density polyethylene carboy that is easy to install. The portable design makes the carboy ideal for transporting distilled water to different lab sites. Included with the reservoir is a spigot for easyMore Product Information