Oxoid CampyGen Compact
Supplier: HARDY DIAGNOSTICS CA
CN020C AG020C
CA10790-378EA 254.96 CAD
CA10790-378 CA76049-416
Oxoid CampyGen Compact
Gas Detection Systems
Simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
The Oxoid CampyGen and CampyGen Compact are simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
Like the AnaeroGen, the CampyGen also requires no catalyst and no water activation steps. No dangerous gas is produced. The final atmosphere is reduced to 6% oxygen, and supplemented with 14% carbon dioxide which is ideal for growth of Campylobacter and other microaerophilic organisms. Sachet,More Product Information