NanoPhotometer® N60-MOBILE UV/Visible Spectrophotometer for NanoVolume Applications, Implen

Supplier: IMPLEN, INC. CA
N60-MOBILE
CA10770-414EA 27422.07 CAD
CA10770-414
NanoPhotometer® N60-MOBILE UV/Visible Spectrophotometer for NanoVolume Applications, Implen
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Implen has become the leading expert for innovative, high-quality spectroscopy instruments and the NanoPhotometer® is trusted by thousands of researchers worldwide.

  • Preprogrammed applications for (labeled) DNA, RNA and oligos (260 nm)
  • (Labeled) purified proteins and peptides (200 to 280 nm)
  • Protein assays (BCA 562 nm, Bradford 595 nm, Lowry 650 nn)
  • Kinetics (200 to 900 nm)
  • Optical density measurements (600 nm)
  • Manual methods (wave length, wave scan, ratio, concentration, standard curve)
  • Recalibration-free
  • Built-in vortex
  • Built-in battery
  • Optional IQOQ
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Display 1024×600 pixels; glove compatible touchscreen
Absorbance accuracy <1.75 % @ 0.7 A @ 280 nm
Photometric range 0.02 - 330 A
Wavelength accuracy ±0.75 nm
Wavelength reproducibility ±0.2 nm
Compatible with Windows 8, 10 (32 and 64 bit), OS X (Intel x86 and Apple M1), iOS and Android OS
Detection limit Detection range BSA: 0.03 - 478 mg/ml (microvolume)
Detection range dsDNA: 1 - 16500 ng/µl (microvolume)
Detector 4096 CMOS Array
Interfaces 2x USB A, USB B, HDMI, Ethernet, WiFi
Languages English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese
Light source Xenon flash lamp
Memory 32 GB
Reproducibility <0.002 A @ 0 - 0.3 A @ 280 nm
CV <1% @ 0.3 - 1.7 A @ 280 nm (Absorbance)
Response time 2.5 - 4.0 s measurement time
Sample compartment 1 sample capacity
Sample size 0.3 - 2.0 µl
Spectral bandwidth <1.8 nm
Dimensions 20×20 cm
Weight <5.2 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR