Terfu Fume Extractor Arms, Movex
Supplier: Movex
MB1500-50 MTI250 MTI500 MTI750 MTI1000 MTI1500 MTF1000 MVK MBF MTI250 MTI500 MTI750 MTI1000 MTI1500 MVK
89038-612EA 973.33 CAD
89038-612 89038-614 89038-616 89038-618 89038-620 89038-622 89038-624 89038-626 89038-628 89038-630 89038-632 89038-634 89038-636 89038-638 89038-640 89038-642 89038-644 89038-646 89038-648 89038-650 89038-652 89038-654 89038-656 89038-658 89038-660 89038-662 89038-664 89038-666 CA10770-088 CA10770-090 CA10770-100 CA10770-102 CA10770-106 CA10770-112
Terfu Fume Extractor Arms, Movex
Fume Extractors
Fume extractor arms with friction joints are ideal for laboratories and electronics applications
Flexible arm allows one-handed adjustments and stays in position when adjusted. Smooth, anodized aluminum tubes and easily dismantled plastic joints facilitate cleaning. Can be mounted on a countertop, ceiling, or wall. Color: white.
Ordering information: For fume extractor arms and hoods with polypropylene and ESD components, contact your Avantor sales representative.
Order Now
SPECIFICATIONS
Environmentally Preferable