HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Canada
115062 115063 115064 115065 115066 115067
CA10769-420CS 3327.38 CAD
CA10769-420 CA10769-494 CA10769-496 CA10769-498 CA10769-500 CA10769-502
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Gloves
Precision engineered to protect your hands and the product, the HyFlex® 11-318 are ultralight design gloves with medium cut performance.
  • Silicone-free
  • Superior tactile sensitivity
  • Ultimate flexibility
  • Low-linting
Featuring an 18 gauge knit with Dyneema® Diamond, Spandex, Nylon and Carbon to provide ultimate tactility and flexibility.
The ultralight fabric and tailored fit of the Hyflex 11-318 create a bare hand-like sensation of comfort and dexterity when performing tasks that demand high tactility. As the first-to-market glove to provide EN Level 3More Product Information