Anti-GARP Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 7B11]

Supplier: Biolegend
352509
CA10759-636EA 328.73 CAD
CA10759-636
Anti-GARP Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 7B11]
Antibodies
Brilliant Violet 421™ anti-human GARP (LRRC32) [7B11]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GARP
Clonality: Monoclonal
Clone: 7B11
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR