Anti-IL-21R Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 2G1-K12]

Supplier: Biolegend
347809
CA10758-788EA 343.1 CAD
CA10758-788
Anti-IL-21R Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 2G1-K12]
Antibodies
Brilliant Violet 421™ anti-human CD360 (IL-21R) [2G1-K12]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-21R
Clonality: Monoclonal
Clone: 2G1-K12
Conjugation: BrilliantMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR