Anti-CD161 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: HP-3G10]

Supplier: Biolegend
339922 339921
CA10758-600EA 694.17 CAD
CA10758-600 CA10759-526
Anti-CD161 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: HP-3G10]
Antibodies
Brilliant Violet 510™ anti-human CD161 [HP-3G10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: Brilliant Violet® 510
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR