Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8]

Supplier: Biolegend
303721
CA10757-796EA 205.86 CAD
CA10757-796
Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8]
Antibodies
Purified anti-human CD41 (MaxPar® Ready) [HIP8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Cynomolgus, Rhesus; Apps: FC, CyTOF®; Size: 100 μg

This product is suitable for use with the MaxPar® Metal Labeling Kits. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the MaxPar® antibody labeling protocol). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: HIP8
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR