Anti-CD27 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: O323]

Supplier: Biolegend
302827 302828
CA302827-BLEA 333.52 CAD
CA302827-BL CA10757-752
Anti-CD27 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: O323]
Antibodies
Brilliant Violet 650™ anti-human CD27 [O323]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: BrilliantMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR