Halogen Moisture Analyzer HE53, METTLER TOLEDO®
Supplier: Mettler Toledo
30100251
CA10752-438EA 5966.03 CAD
CA10752-438
Halogen Moisture Analyzer HE53, METTLER TOLEDO®
Analysers Moisture Analysers
The HE53 Halogen Moisture Analyzer is a reliable and cost-effective solution for determining moisture content.
  • Reliable and cost-effective
  • Simple to use
  • Three-step process
  • Metal base
  • Stainless steel surface
The instrument is simple to use, thanks to straightforward navigation and on-screen guidance.
Proper installation and initial calibration are the key to ensuring the moisture analyzer performs properly from day one. A METTLER TOLEDO® authorized service technician performs installation qualification and operational qualification (IQ/OQ).