Buffer No. 16, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
U558
CA10693-110PK 4438.85 CAD
CA10693-110
Buffer No. 16, Hardy Diagnostics
Buffers Cell Culture Buffers
For the use in determination of antimicrobial activity.
For the use in determination of antimicrobial activity. Formulated according to USP <81>.
MINIMUM ORDER: 5 packs