Anti-FasL (CD178.1) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Kay-10]

Supplier: Biolegend
106805
CA106805-BLEA 205.86 CAD
CA106805-BL
Anti-FasL (CD178.1) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Kay-10]
Antibodies
PE anti-mouse CD178.1 (FasL) [Kay-10]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: FasL (CD178.1)
Clonality: Monoclonal
Clone: Kay-10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR