HistoTainer™ Prefilled Specimen Containers, with 10% Neutral Buffered Formalin, Simport Scientific
Supplier: Simport Scientific
M961-20FW M961-40FW M961-60FW M961-90FW M961-120FW
CA97060-890CS 136.68 CAD
CA97060-890 CA97060-892 CA97060-894 CA97060-896 CA97060-898 89499-250 89499-252 89499-254 89499-256 89499-258 CA10662-320 CA10662-322 CA10662-324 CA10662-326 CA10662-328
HistoTainer™ Prefilled Specimen Containers, with 10% Neutral Buffered Formalin, Simport Scientific
Sample Containers General Purpose Containers
Shatter resistant and leak proof polypropylene containers are designed for collection, transport, and storage of histology specimens
The HistoTainer™ I includes a tamper evident leak-proof screw cap. A special push-tab mechanism is integrated in the container. These containers may also be used without using the tamper evident locking mechanism. The HistoTainer™ II includes a regular leak-proof screw cap.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used in manufacture are free from latex. Containers are 95kPa compliant.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable