ChillBlock™ Thermo-Conductive Racks, Simport Scientific
Supplier: Simport Scientific
S700-35 S700-50 S700-6 S700-8 S700-10 S700-14 S700-56 S700-80 S700-16 S700-1 S700-4 S700-35 S700-50 S700-6 S700-8 S700-10 S700-14 S700-56 S700-80 S700-16 S700-90 S700-1 S700-4
10662-278EA 823.59 CAD
10662-278 10662-280 10662-282 10662-284 10662-292 10662-294 10662-296 10662-298 10662-300 10662-302 10662-304 10662-306 10662-334 10662-336 10662-338 10662-342 10662-344 10662-442 10662-444 CA10662-278 CA10662-280 CA10662-282 CA10662-284 CA10662-292 CA10662-294 CA10662-296 CA10662-298 CA10662-300 CA10662-302 CA10662-304 CA10662-306 CA10662-334 CA10662-336 CA10662-338 CA10662-340 CA10662-342 CA10662-344 CA10662-346 CA10662-442 CA10662-444
ChillBlock™ Thermo-Conductive Racks, Simport Scientific
Racks Tube Racks
ChillBlock™ thermo-conductive metal alloy tube racks eliminate inconsistencies that result from inserting tubes directly into ice, dry ice, alcohol baths, water baths, and other common laboratory temperature sources
ChillBlock™ tube racks are available in a variety of sizes for vessels such as microcentrifuge tubes, cryogenic vials, PCR tubes, SBS-compliant strips and plates, and 15mL and 50mL tubes.
Racks adapt from ambient to <4°C in 60 to 90 seconds on ice; from ambient to –78°C in 5 to 7 minutes on dry ice; and from ambient to –140°C in 15 minutes when used above liquid nitrogen.More Product Information