TitraLab® AT1000 Potentiometric Titrator, 1 Burette, 2 Pumps
Supplier: Hach
AP0007.AT1122 AP0012.AT1122 AT1122.97
10662-156EA 3312.98 CAD
10662-156 10662-202 10662-256 CA10662-156 CA10662-202 CA10662-256
TitraLab® AT1000 Potentiometric Titrator, 1 Burette, 2 Pumps
Titrators
Automatic titration without all the complications.
  • Reliable titration results
  • Eliminates operator interpretation and manual processes
  • Simple setup and titration
  • Application-specific functions
Simplify complex titration with the TitraLab® AT1000 from Hach.
Eliminates operator interpretation and manual processes with automatic titration to quickly deliver accurate and repeatable results. Pre-programmed titration methods detect end points and eliminate manual calculations to make results easier to achieve without advanced programming. Application-More Product Information