TitraLab® AT1000 Series Potentiometric Titrator, 1 Burette, Hach
Supplier: Hach
AP0008.AT1102 AP0015.AT1102 AP0016.AT1102 AP0006.AT1102 AT1102.97 AP0001.AT1102 AP0002.AT1102
10662-158EA 3547.68 CAD
10662-158 10662-164 10662-166 10662-174 10662-242 10662-252 10662-262 10662-264 CA10662-158 CA10662-164 CA10662-166 CA10662-174 CA10662-252 CA10662-262 CA10662-264
TitraLab® AT1000 Series Potentiometric Titrator, 1 Burette, Hach
Titrators
Automatic titration without all the complications.
  • Reliable titration results
  • Eliminates operator interpretation and manual processes
  • Simple setup and titration
  • Application-specific functions
Simplify complex titration with the TitraLab AT1000 from Hach.
Eliminates operator interpretation and manual processes with automatic titration to quickly deliver accurate and repeatable results. Pre-programmed titration methods detect end points and eliminate manual calculations to make results easier to achieve without advanced programming. Application-More Product Information