Anti-CD102 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 3C4 (MIC2/4)]

Supplier: Biolegend
105609
CA105609-BLEA 161.18 CAD
CA105609-BL
Anti-CD102 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 3C4 (MIC2/4)]
Antibodies
Alexa Fluor® 488 anti-mouse CD102 [3C4 (MIC2/4)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD102
Clonality: Monoclonal
Clone: 3C4 (MIC2/4)
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR