Affinity Chromatography Columns, Tagged Package, His, Cytiva
Supplier: Cytiva
29-0588-03
CA10498-068EA 336.84 CAD
CA10498-068
Affinity Chromatography Columns, Tagged Package, His, Cytiva
Chromatography Columns
His package is designed for the convenient purification of histidine-tagged proteins.
The His package comes with ready-to-use columns packed with high performance media developed for high yield (HisTrap™ HP, 1×1 ml) or high purity (HiTrap™ TALON crude, 1×1 ml) and a HiTrap™ desalting column (1×5 ml) for removing imidazole from eluate.
Delivery information: The kit contains one of each HisTrap™ HP (1×1 ml), HiTrap™ TALON crude (1×1 ml) and HiTrap™ Desalting (1×5 ml) columns.