Inlet Filter Assembly Kit, Cytiva
Supplier: Cytiva
18-1113-15
CA10497-990EA 615.11 CAD
CA10497-990
Inlet Filter Assembly Kit, Cytiva
Filters Liquid Process Filters
Kit contents include one support net, one filter (mounted on fliterholder), and one screw lid and cap membrane.
The wetted parts, except for the membrane, are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, with the exception of 100% Ethyl acetate, 100% Hexane, and 100% Tetrahydrofuran.