Anti-CD49e Armenian Hamster Monoclonal Antibody (Biotin) [clone: HMalpha5-1]

Supplier: Biolegend
103904
CA103904-BLEA 430.87 CAD
CA103904-BL
Anti-CD49e Armenian Hamster Monoclonal Antibody (Biotin) [clone: HMalpha5-1]
Antibodies
Biotin anti-mouse/rat CD49e [HMα5-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49e
Clonality: Monoclonal
Clone: HMalpha5-1
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR