EnGen Mutation Detection Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E3321S
CA103049-174EA 358 CAD
CA103049-174
EnGen Mutation Detection Kit, New England Biolabs
Electrophoresis Systems Mutation Detection Systems
The EnGen Mutation Detection Kit provides reagents for detection of on-target genome editing events.
In the first step, targeted regions from cells whose genomes were targeted (i.e. CRISPR/Cas9, TALENs, Zinc-finger Nucleases) are amplified using Q5 Hot Start High-Fidelity 2X Master Mix. Upon denaturation and re-annealing, heteroduplexes are formed when mutations from insertions and deletions (indels) are present in the amplicon pool. In the second step, annealed PCR products are digested with EnGen T7 Endonuclease I, a structure-specific enzyme that will recognize mismatches larger than 1More Product Information