NEBNEXT® Small RNA Library Prep Set for Illumina (Multiplex Compatible), New England Biolabs
Supplier: New England Biolabs (NEB)
E7330L E7330S
CA102500-112EA 6637 CAD
CA102500-112 CA102500-114
NEBNEXT® Small RNA Library Prep Set for Illumina (Multiplex Compatible), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The novel NEBNext® Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries
The NEBNext® Small RNA Library Prep Set for Illumina® (Multiplex Compatible) enables use with your own barcode system. The kit contains adaptors but does not include multiplex primers.