Chromatography Columns, Spherical Joint, Fritted Disc, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0195261C CS-C0195262C CS-C0195263C CS-C0195264C CS-C0195321C CS-C0195322C CS-C0195323C CS-C0195324C CS-C0195461C CS-C0195462C CS-C0195463C CS-C0195464C CS-C0195601C CS-C0195602C CS-C0195603C CS-C0195604C CS-C0195605C CS-C0195801C CS-C0195802C CS-C0195803C CS-C0195804C CS-C0195805C CS-C0195171C CS-C0195172C CS-C0195173C
CA10201-092CS 716.11 CAD
CA10201-092 CA10201-094 CA10201-096 CA10201-098 CA10201-100 CA10201-102 CA10201-104 CA10201-106 CA10201-108 CA10201-110 CA10201-112 CA10201-114 CA10201-116 CA10201-118 CA10201-120 CA10201-122 CA10201-124 CA10201-126 CA10201-128 CA10201-130 CA10201-132 CA10201-134 CA10203-554 CA10203-556 CA10203-558
Chromatography Columns, Spherical Joint, Fritted Disc, ChemScience
Chromatography Columns
These columns have a Teflon® stopcock and a spherical joint.
They are equipped with a coarse porosity sealed in fritted disc to support column packing.
The spherical joint used with pinch clamp makes quick and tight connection under pressure for flash chromatography.