Chromatography Columns, Spherical Joint, Fritted Disc, ChemScience
Supplier: CHEM SCIENCE INC CA
CS-C0195261C CS-C0195262C CS-C0195263C CS-C0195264C CS-C0195321C CS-C0195322C CS-C0195323C CS-C0195324C CS-C0195461C CS-C0195462C CS-C0195463C CS-C0195464C CS-C0195601C CS-C0195602C CS-C0195603C CS-C0195604C CS-C0195605C CS-C0195801C CS-C0195802C CS-C0195803C CS-C0195804C CS-C0195805C CS-C0195171C CS-C0195172C CS-C0195173C
CA10201-092CS 716.11 CAD
CA10201-092 CA10201-094 CA10201-096 CA10201-098 CA10201-100 CA10201-102 CA10201-104 CA10201-106 CA10201-108 CA10201-110 CA10201-112 CA10201-114 CA10201-116 CA10201-118 CA10201-120 CA10201-122 CA10201-124 CA10201-126 CA10201-128 CA10201-130 CA10201-132 CA10201-134 CA10203-554 CA10203-556 CA10203-558
Chromatography Columns, Spherical Joint, Fritted Disc, ChemScience
Chromatography Columns
These columns have a Teflon® stopcock and a spherical joint.
They are equipped with a coarse porosity sealed in fritted disc to support column packing.
The spherical joint used with pinch clamp makes quick and tight connection under pressure for flash chromatography.
Order Now
SPECIFICATIONS
Environmentally Preferable