SphI-HF®, Restriction Enzymes, New England Biolabs

Supplier: New England Biolabs (NEB)

R3182M
CA101641-168EA 458 CAD
CA101641-168
SphI-HF®, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes

An E.coli strain that carries the cloned and modified SphI gene from Streptomyces phaeochromogenes (NRRL B-3559).


High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name SphI-HF
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR