NEBNEXT® mRNA Second Strand Synthesis Module, New England Biolabs
Supplier: New England Biolabs (NEB)
B6117S E6111L E6111S
CA101640-776EA 54 CAD
CA101640-776 CA101640-872 CA101640-874
NEBNEXT® mRNA Second Strand Synthesis Module, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Second Strand Synthesis Module has been optimized to generate double stranded cDNA from first strand cDNA
  • NEBNext Second Strand Synthesis (dNTP-free) reaction buffer
The NEBNext Second Strand Synthesis Module is provided as a master mix to maximize efficiency and convenience in RNA sample preparation workflows.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more