SENSATION® Powder free Examination Gloves, Microflex®, Ansell
Supplier: Microflex
N730 N731 N732 N733 N734
CA10159-898BX 30.45 CAD
CA10159-898 CA10159-900 CA10159-902 CA10159-904 CA10159-906
SENSATION® Powder free Examination Gloves, Microflex®, Ansell
Gloves
Powder-free, blue-colored examination gloves with enhanced comfort and tactile sensitivity.
  • Gloves are nonsterile and latex-free
  • Powder-free
  • Ambidextrous
  • Beaded cuffs
  • Disposable nitrile goves
Microflex® Sensation® N73 powder-free examination gloves provide exceptional fit with textured fingertips for an excellent grip for use with small instruments. FDA cleared 'Low Dermatitis' claim, which means Sensation® gloves have been clinically shown to reduce incidence of the redness and itching associated with contact dermatitis.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Powdered/Powder-free Powder-free
Cuff style Beaded
Cuff thickness 2.0 mil
Finger thickness 3.9 mil
Palm thickness 2.4 mil
Tensile strength 29 MPa