Rose Bengal Agar with Chloramphenicol 1.5X
Supplier: HARDY DIAGNOSTICS CA
Q81
CA10158-952PK 780.43 CAD
CA10158-952
Rose Bengal Agar with Chloramphenicol 1.5X
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and enumeration of fungi from environmental and food sources.
For the selective isolation and enumeration of fungi from environmental and food sources.