ACTERO™ Salmonella Enrichment Media, FoodChek Systems
Supplier: FOODCHEK SYSTEMS INC. CA
FCM-009 FCM-049 FCM-050 FCM-051 FCM-085 FCM-086 FCM-087 FCM-088 FCM-089 FCM-090 FCM-064 FCM-065 FCM-066 FCM-144 FCM-145 FCM-146 FCM-147 FCM-148 FCM-140 FCM-141 FCM-142 FCM-143 FCM-188 FCM-189 FCM-190
CA97068-836EA 921.92 CAD
CA97068-836 CA10066-670 CA10066-672 CA10066-674 CA10066-792 CA10066-794 CA10066-796 CA10066-798 CA10066-800 CA10066-802 CA10066-868 CA10066-870 CA10066-872 CA10158-444 CA10158-446 CA10158-448 CA10158-450 CA10158-452 CA10158-516 CA10158-518 CA10158-520 CA10158-522 CA10158-922 CA10158-924 CA10158-926
ACTERO™ Salmonella Enrichment Media, FoodChek Systems
Media for Microbiology Dehydrated Media for Microbiology
Food and environmental samples require enrichment before pathogen testing enabling target pathogen present to grow and be detected in test samples
Using Actero™ Elite Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system," such as lateral flow, ELISA and PCR platforms.
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.
The patented Actero™ Elite Enrichment Media are enhanced formulations that reduce the enrichment time required for accurate pathogen testing. Actero™ Salmonella Enrichement Media recieved the AOAC-PTM (041403) accreditation to be used as single-step recover and enrichment of Salmonella spp. from selected food and environmental samples.