HiScreen Capto S ImpAct Columns, Cytiva

Supplier: Cytiva
17-3717-47
CA10158-354EA 425.19 CAD
CA10158-354
HiScreen Capto S ImpAct Columns, Cytiva
Chromatography Columns
HiScreen™ Capto™ S ImpAct columns are prepacked with Capto™ S ImpAct cation exchange chromatography medium.

  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Easily connected in series to achieve 200 mm bed height
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Strong cation exchange BioProcess medium, designed for immediate purification and polishing of a wide range of biomolecules, especially monoclonal antibodies
  • Flow rate of 1.2 to 2.3 ml/min

More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Binding capacity > 85 mg BSA/mL medium, > 90 mg lysozyme/mL medium, > 100 mg IgG/ mL medium
pH range 4 - 12
Storage 4 to 30 °C, 20% Ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR