Sera-Mag SpeedBeads® Neutravidin Magnetic Beads, Cytiva
Supplier: Cytiva
78152110150 78152110350 78152104011150
CA10146-084EA 2791.63 CAD
CA10146-084 CA10146-086 CA10146-088
Sera-Mag SpeedBeads® Neutravidin Magnetic Beads, Cytiva
Beads Magnetic Beads
Sera-Mag SpeedBeads Neutravidin-Coated Magnetic Particles exhibit high affinity for biotinylated ligands such as proteins, nucleic acids, and peptides.
  • Even coating of Neutravidin protein provides reliable results
  • Low non-specific binding for fast and clean purification
  • Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
  • Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
  • Uniform, nominal 1 μm diameter provides excellent lot-to-
More Product Information
Order Now
SPECIFICATIONS
Binding capacity 3500-4500 pg/mg beads
Environmentally Preferable
Particle size nominal 1 µm diameter