Hei-VAP Rotary Evaporator RPM Regulated Vacuum Pumps
Supplier: Heidolph NA, LLC
036308100CA
CA10145-674EA 12719.23 CAD
CA10145-674
Hei-VAP Rotary Evaporator RPM Regulated Vacuum Pumps
Pumps Vacuum Pumps
Combinable with a condenser.
Hei-VAC Vario Control
For Hei-VAP Expert Control / Hei-VAP Ultimate Control. Even if the gas ballast valve is open an excellent ultimate vacuum is reached when working with easily condensable vapors. This makes distilling high boiling point solvents such as DMF or DMSO possible at low bath temperatures.
Hei-VAC Vario Station
For Hei-VAP Expert Control / Hei-VAP Ultimate Control. Fully controllable stand-alone pumping unit including vacuum controller. Precise vacuum control avoids bumping and eliminates foaming of your evaporation solution. Due to automaticMore Product Information