Hemo KlenTaq, New England Biolabs
Supplier: New England Biolabs (NEB)
M0332L M0332S
CA101449-330EA 758 CAD
CA101449-330 CA101449-332
Hemo KlenTaq, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hemo KlenTaq is a truncated version of Taq DNA Polymerase, lacking the first 280 amino acids
Hemo KlenTaq also contains mutations that make it resistant to inhibitors present in whole blood. It can amplify DNA from whole blood samples from humans and mice, without the need for DNA extraction. Hemo KlenTaq tolerates up to 20% whole blood in a 25 µl reaction (30% in a 50 µl reaction). Hemo KlenTaq works well with most common anticoagulants, including heparin, citrate and EDTA.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more