LONGAMP® Taq PCR Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E5200S
CA101444-720EA 204 CAD
CA101444-720
LONGAMP® Taq PCR Kit, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The LongAmp Taq PCR Kit features LongAmp Taq DNA Polymerase, which is a unique blend of Taq and Deep VentR ™ DNA Polymerases
The 3´→5´ exonuclease activity of Deep VentR Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq Polymerase alone. The LongAmp Taq PCR Kit is supplied with LongAmp Taq Reaction Buffer, which is optimized to give highest PCR yields up to 30 kb. The kit can generate a wide range of PCR products, up to 30 kb from plasmid or from human genomic DNA.