Nickel sulfate 0.1 M, sterile

Supplier: TEKNOVA CA

N6565
CA101320-430EA 468.56 CAD
CA101320-430
Nickel sulfate 0.1 M, sterile
Nickel sulphate

Nickel sulfate solution, for use in Histidine-tagged proteins purifications procedures.

Formula: NiO₄S
MDL Number: MFCD00011146
CAS Number: 7786-81-4
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR