AsiSI

Supplier: New England Biolabs (NEB)

R0630L R0630S
CA101229-490EA 480 CAD
CA101229-490 CA101229-492
AsiSI
Nucleic Acid Reagents Restriction Enzymes

An E.coli strain that carries the cloned AsiSI gene from Arthrobacter species (S.K. Degtyarev).

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name AsiSI
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR