Lambda DNA-BstE II Digest, New England Biolabs
Supplier: New England Biolabs (NEB)
N3014S
CA101228-444EA 127 CAD
CA101228-444
Lambda DNA-BstE II Digest, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
The BstEII digest of lambda DNA (cI857 ind 1 Sam 7) yields 14 fragments suitable for use as molecular weight standards for agarose gel electrophoresis
The approximate mass of DNA in each of the bands is provided (assuming a 1.0 μg load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.