Endouclease III (Nth)
Supplier: New England Biolabs (NEB)
M0268S
CA101228-320EA 124 CAD
CA101228-320
Endouclease III (Nth)
Enzymes
Endonuclease III (Nth) protein from E.coli acts as both N-glycosylase and a AP-lyase.
The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating a basic (AP site). The AP-lyase activity of the enzyme cleaves 3´ to the AP site leaving a 5´ phosphate and a 3´ ring opened sugar. Some of the damaged bases recognized and removed by Endouclease III include urea, 5, 6 dihydroxythymine, thymine glycol, 5-hydroxy-5 methylhydanton, uracil glycol, 6-hydroxy-5, 6-dihdrothimine and methyltartronylurea.