Optifix® Dispensers, MilliporeSigma
Supplier: MilliporeSigma
10108044-1 10108048-1 10108058-1 10108133-1 10108144-1 10108153-1 10110638-1 10110644-1 108056G-1 10110738-1 10110748-1 10108027-1 10108033-1 10108051-1 10108127-1 10108138-1 10108148-1 108056-1 108053-1 10110727-1
CA10108044-1EA 2485 CAD
CA10108044-1 CA10108048-1 CA10108058-1 CA10108133-1 CA10108144-1 CA10108153-1 CA10110638-1 CA10110644-1 CA108056G-1 CAM10110738-1 CAM10110748-1 CA10108027-1 CA10108033-1 CA10108038-1 CA10108051-1 CA10108127-1 CA10108138-1 CA10108148-1 CA108056-01 CAE18053-01 CAM10110727-1
Optifix® Dispensers, MilliporeSigma
Bottle Top Dispensers
Optifix® Dispensers are available in four models, each of which is designed to increase efficiency and provide additional safety to daily laboratory operations. These dispensers are manufactured to the highest quality standards, ensuring accurate, reliable, and easy dispensing.
Ideal for critical dispensing applications, the Optifix® BASIC dispenser is used for aqueous solutions, light acids, and crystallizing substances
The Optifix® SOLVENT dispenser is useful for all organic solvents and materials that swell PTFE. The piston is glass. The Optifix® SOLVENT dispenser with vapor trap isMore Product Information