MsGM™ Mesangial Cell Growth Medium
Supplier: LONZA PHARMA - BIOSCIENCE CA
CC-4146 CC-3146 CC-3147
CA10063-940EA 432.53 CAD
CA10063-940 CA10065-256 CA10065-258
MsGM™ Mesangial Cell Growth Medium
Cell Culture Media
MsGM™ Mesangial Cell Growth Medium; is formulated for the growth of renal mesangial cells. MsGM™ contains 5% serum.
Delivery information: Kit includes MsBM basal medium 500 mL and MsGM SingleQuot kit with supplement and growth factors,1×Red cap vial with GA-1000 0.50 ml, 1×bottle FBS 25.00 ml.
Order Now
SPECIFICATIONS
Environmentally Preferable